Learn More
Biolegend Brilliant Violet 650™ anti-mouse IL-17A
Supplier:  Biolegend 506929
Encompass_Preferred
Brilliant Violet 650™ anti-mouse IL-17A [TC11-18H10.1]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 125 uL
Catalog No. 50-403-675