Learn More
Biolegend Brilliant Violet 650™ anti-mouse Ly-6G/Ly-6C (Gr-1)
Supplier:  Biolegend 108441
Encompass_Preferred
Brilliant Violet 650™ anti-mouse Ly-6G/Ly-6C (Gr-1) [RB6-8C5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 125 uL
Catalog No. 50-605-966