Learn More
Biolegend Brilliant Violet 650™ Rat IgG2b, κ Isotype Ctrl
Supplier:  Biolegend 400651
Encompass_Preferred
Brilliant Violet 650™ Rat IgG2b, κ Isotype Ctrl [RTK4530]; Isotype: Rat IgG2b, κ; Apps: FC, ICFC; Size: 50 ug
Catalog No. 50-605-405