Learn More
Biolegend Brilliant Violet 711™ anti-human CD19
Supplier:  Biolegend 302245
Encompass_Preferred
Brilliant Violet 711™ anti-human CD19 [HIB19]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-604-005