Learn More
Biolegend Brilliant Violet 711™ anti-human CD206 (MMR)
Supplier:  Biolegend 321135
Encompass_Preferred
Brilliant Violet 711™ anti-human CD206 (MMR) [15-2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-3851