Learn More
Biolegend Brilliant Violet 711™ anti-human CD31
Supplier:  Biolegend 303135
Encompass_Preferred
Brilliant Violet 711™ anti-human CD31 [WM59]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-207-2951