Learn More
Biolegend Brilliant Violet 711™ anti-human CD45RO
Supplier:  Biolegend 304235
Encompass_Preferred
Brilliant Violet 711™ anti-human CD45RO [UCHL1]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Chimpanzee, Cynomolgus, Common Marmoset; Apps: FC; Size: 25 tests
Catalog No. 50-403-245