Learn More
Biolegend Brilliant Violet 711™ anti-human TNF-α
Supplier:  Biolegend 502939
Encompass_Preferred
Brilliant Violet 711™ anti-human TNF-α [MAb11]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cat (Feline), Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Swine (Pig, Porcine), Pigtailed Macaque, Sooty Mangabey; Apps: ICFC, FC; Size: 25 tests
Catalog No. 50-208-5582