Learn More
Biolegend Brilliant Violet 711™ anti-human CD56 (NCAM)
Supplier:  Biolegend 362541
Encompass_Preferred
Brilliant Violet 711™ anti-human CD56 (NCAM) [5.1H11]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-8060