Learn More
Biolegend Brilliant Violet 711™ anti-human CD62L
Supplier:  Biolegend 304860
Encompass_Preferred
Brilliant Violet 711™ anti-human CD62L [DREG-56]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cattle (Bovine, Cow); Apps: FC; Size: 100 tests
Catalog No. 50-207-3139