Learn More
Biolegend Brilliant Violet 711™ anti-mouse CD184 (CXCR4)
Supplier:  Biolegend 146517
Encompass_Preferred
Brilliant Violet 711™ anti-mouse CD184 (CXCR4) [L276F12]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC1547162