Learn More
Biolegend Brilliant Violet 711™ anti-mouse CD274 (B7-H1, PD-L1)
Supplier:  Biolegend 124319
Encompass_Preferred
Brilliant Violet 711™ anti-mouse CD274 (B7-H1, PD-L1) [10F.9G2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-850-339