Learn More
Biolegend Brilliant Violet 711™ anti-mouse CD64 (FcγRI)
Supplier:  Biolegend 139311
Encompass_Preferred
Brilliant Violet 711™ anti-mouse CD64 (FcγRI) [X54-5/7.1]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-711-882