Learn More
Biolegend Brilliant Violet 711™ anti-mouse IFN-γ
Supplier:  Biolegend 505835
Encompass_Preferred
Brilliant Violet 711™ anti-mouse IFN-γ [XMG1.2]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 125 uL
Catalog No. 50-403-668