Learn More
Biolegend Brilliant Violet 711™ Mouse IgG1, κ isotype Ctrl
Supplier:  Biolegend 400168
Encompass_Preferred
Brilliant Violet 711™ Mouse IgG1, κ isotype Ctrl [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse ; Apps: FC, ICFC; Size: 100 tests
Catalog No. 50-850-899