Learn More
Biolegend Brilliant Violet 785™ anti-human CD19
Supplier:  Biolegend 363028
Encompass_Preferred
Brilliant Violet 785™ anti-human CD19 [SJ25C1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-850-877