Learn More
Biolegend Brilliant Violet 785™ anti-human CD27
Supplier:  Biolegend 302832
Encompass_Preferred
Brilliant Violet 785™ anti-human CD27 [O323]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests
Catalog No. 50-604-013