Learn More
Biolegend Brilliant Violet 785™ anti-human CD8a
Supplier:  Biolegend 301045
Encompass_Preferred
Brilliant Violet 785™ anti-human CD8a [RPA-T8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Pigtailed Macaque, Sooty Mangabey; Apps: FC; Size: 25 tests
Catalog No. 50-403-188