Learn More
Biolegend Brilliant Violet 785™ anti-human HLA-DR
Supplier:  Biolegend 307641
Encompass_Preferred
Brilliant Violet 785™ anti-human HLA-DR [L243]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Dog (Canine), Marmoset, Rhesus, Squirrel Monkey, Tamarin; Apps: FC; Size: 25 tests
Catalog No. 50-403-262