Learn More
Biolegend Brilliant Violet 785™ anti-human CD107a (LAMP-1)
Supplier:  Biolegend 328644
Encompass_Preferred
Brilliant Violet 785™ anti-human CD107a (LAMP-1) [H4A3]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1116453