Learn More
Biolegend Brilliant Violet 785™ anti-human CD33
Supplier:  Biolegend 303428
Encompass_Preferred
Brilliant Violet 785™ anti-human CD33 [WM53]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee; Apps: FC; Size: 100 tests
Catalog No. NC1451261