Learn More
Biolegend Brilliant Violet 785™ anti-mouse CD16.2 (FcγRIV)
Supplier:  Biolegend 149535
Encompass_Preferred
Brilliant Violet 785™ anti-mouse CD16.2 (FcγRIV) [9E9]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-207-1884