Learn More
Biolegend Brilliant Violet 785™ anti-mouse CD4
Supplier:  Biolegend 100551
Encompass_Preferred
Brilliant Violet 785™ anti-mouse CD4 [RM4-5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 125 uL
Catalog No. 50-402-926