Learn More
Biolegend Brilliant Violet 785™ anti-mouse CD45
Supplier:  Biolegend 103149
Encompass_Preferred
Brilliant Violet 785™ anti-mouse CD45 [30-F11]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-103-0520