Learn More
Biolegend Brilliant Violet 785™ Mouse IgG1, κ isotype Ctrl
Supplier:  Biolegend 400169
Encompass_Preferred
Brilliant Violet 785™ Mouse IgG1, κ isotype Ctrl [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse ; Apps: FC, ICFC; Size: 25 tests
Catalog No. NC1631681