Learn More
Medchemexpress LLC BVDU 5′-triphosphate ammonium | 98.0% | 590.11 g/mol | C11H19BrN3O14P3 | 10mg
SDP
Catalog No. 5000268290
Encompass_Preferred
BVDU 5 -Triphosphate ammonium is an antivirus agent with 5 -Triphosphate label targeting viral DNA polymerase BVDU 5 -Triphosphate ammonium shows excellent selectivity against varicella-zoster virus (VZV) and herpes simplex virus type 1 (HSV-1) due to a specific phosphorylation by the virus-encoded thymidine kinase
Catalog No. 50-002-68290 Supplier Medchemexpress LLC Supplier No. HYW747737A10MG
BVDU 5 -Triphosphate ammonium is an antivirus agent with 5 -Triphosphate label targeting viral DNA polymerase BVDU 5 -Triphosphate ammonium shows excellent selectivity against varicella-zoster virus (VZV) and herpes simplex virus type 1 (HSV-1) due to a specific phosphorylation by the virus-encoded thymidine kinase