Learn More
Aviva Systems Biology Corp BVES Antibody - middle region (ARP46823_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46823_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BVES. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BVES. Alternate names: HBVES, MGC42413, POP1, POPDC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-54015