Learn More
Vici Precision Sampling, Inc. BVLD NEEDLE FOR A-2 SYRNG 3PK
Catalog No. NC9496851
Encompass_Preferred
Needles Gas A-2, C & D .029 in. x .012 in. x 2 in. (22 gauge) bevel open end pkg./3
Catalog No. NC9496851 Supplier Vici Precision Sampling, Inc. Supplier No. 943051
Needles Gas A-2, C & D .029 in. x .012 in. x 2 in. (22 gauge) bevel open end pkg./3