Learn More
Enzo Life Sciences Biliverdin Reductase (rat), (recombinant) (200µg)
SDP
Supplier:  Enzo Life Sciences ADIOSP450F
Encompass_Preferred
Alternative name: BLVR. Purity: ≥95% (SDS-PAGE; Western blot). MW: ~33kDa. Formulation: Liquid. In 50mM TRIS-HCl, pH 8.0, containing 1.0mM EDTA, and 5.0mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB Activity assay.
Catalog No. 50-200-7380