Learn More
Medchemexpress LLC HY-18055 5mg Medchemexpress, BVT-14225 CAS:376638-65-2 Purity:>98%
SDP
Catalog No. 502082015
Encompass_Preferred
Medchemexpress, HY-18055 5mg BVT-14225 CAS:376638-65-2 BVT-14225 is a selective 11β-Hydroxysteroid dehydrogenase type 1 (11β-HSD1) inhibitor with an IC50 of 52 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2015 Supplier Medchemexpress LLC Supplier No. HY180555MG
Medchemexpress, HY-18055 5mg BVT-14225 CAS:376638-65-2 BVT-14225 is a selective 11β-Hydroxysteroid dehydrogenase type 1 (11β-HSD1) inhibitor with an IC50 of 52 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.