Learn More
Medchemexpress LLC HY-18054 5mg Medchemexpress, BVT 2733 CAS:376640-41-4 Purity:>98%
SDP
Catalog No. 502082014
Encompass_Preferred
Medchemexpress, HY-18054 5mg BVT 2733 CAS:376640-41-4 BVT 2733 is a potent, selective, and orally active non-steroidal 11β-hydroxydehydrogenase 1 (11β-HSD1) inhibitor. BVT 2733 is potently against the mouse enzyme (IC50=96 nM) over the human enzyme (IC50=3341 nM). BVT 2733 has the potential for the study of arthritis and obesity related disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2014 Supplier Medchemexpress LLC Supplier No. HY180545MG
Medchemexpress, HY-18054 5mg BVT 2733 CAS:376640-41-4 BVT 2733 is a potent, selective, and orally active non-steroidal 11β-hydroxydehydrogenase 1 (11β-HSD1) inhibitor. BVT 2733 is potently against the mouse enzyme (IC50=96 nM) over the human enzyme (IC50=3341 nM). BVT 2733 has the potential for the study of arthritis and obesity related disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.