Learn More
Abcam BVT 948, Protein tyrosine phosphatase (PTP) inhibitor, 50MG
Catalog No. 502706699
Encompass_Preferred
MW 241.24 Da, Purity >99%. Non-competitive, irreversible protein tyrosine phosphatase (PTP) inhibitor (IC₅₀ values are 0.9 (PTP1B), 1.7 (TCPTP), 0.09 (SHP-2), 1.5 (LAR) and 0.7 (YopH) μM). Active in vivo and cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6699 Supplier Abcam Supplier No. AB14130450MG
MW 241.24 Da, Purity >99%. Non-competitive, irreversible protein tyrosine phosphatase (PTP) inhibitor (IC₅₀ values are 0.9 (PTP1B), 1.7 (TCPTP), 0.09 (SHP-2), 1.5 (LAR) and 0.7 (YopH) μM). Active in vivo and cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement