Learn More
Medchemexpress LLC BW-723C86 | 160521-72-2 | MFCD01321066 | 99.8% | 322.85 | C16H19ClN2OS | 50 MG
SDP
Catalog No. 5000212886
Encompass_Preferred
BW-723C86 is an orally active, selective 5-HT2B receptor agonist supplied for research use in preclinical pharmacology. It exhibits anxiolytic-like effects and other physiological actions in animal studies and is provided as a high-purity solid with specified storage recommendations to preserve stability.
  • Selective 5-HT2B receptor agonist suitable for receptor studies.
  • High purity (99.8%).
  • Orally active in preclinical models.
  • Solid form, white to light brown, facilitating handling and formulation.
  • Defined storage conditions to maintain stability (sealed, away from moisture).
Catalog No. 50-002-12886 Supplier Medchemexpress LLC Supplier No. HY10136950MG
BW-723C86 is an orally active, selective 5-HT2B receptor agonist supplied for research use in preclinical pharmacology. It exhibits anxiolytic-like effects and other physiological actions in animal studies and is provided as a high-purity solid with specified storage recommendations to preserve stability.
  • Selective 5-HT2B receptor agonist suitable for receptor studies.
  • High purity (99.8%).
  • Orally active in preclinical models.
  • Solid form, white to light brown, facilitating handling and formulation.
  • Defined storage conditions to maintain stability (sealed, away from moisture).