Learn More
MSC BW Technologies by Honeywell GasAlertQuattro Alkaline 4-Gas Detector, 1 ea
Catalog No. 5000781411
Encompass
The GasAlertQuattro is a rugged and reliable four-gas detector designed for industrial applications like confined space entry. It features simple one-button operation and flexible power options, with a graphic LCD displaying operational information. IntelliFlash provides continuous visual confirmation of compliance, and it is fully compatible with the MicroDock II automatic test and calibration system.
  • Rugged and reliable design
  • Four-gas detection (%LEL, O2, H2S, CO)
  • Simple one-button operation
  • Flexible power options
  • Graphic LCD with easy-to-identify icons
  • Displays operational information, bump test, and calibration status
  • IntelliFlash for continuous visual confirmation of operation and compliance
  • Suitable for confined space entry
  • Compatible with MicroDock II automatic test and calibration system
  • Water resistant
Catalog No. 50-007-81411 Supplier MSC Supplier No. 50006048
The GasAlertQuattro is a rugged and reliable four-gas detector designed for industrial applications like confined space entry. It features simple one-button operation and flexible power options, with a graphic LCD displaying operational information. IntelliFlash provides continuous visual confirmation of compliance, and it is fully compatible with the MicroDock II automatic test and calibration system.
  • Rugged and reliable design
  • Four-gas detection (%LEL, O2, H2S, CO)
  • Simple one-button operation
  • Flexible power options
  • Graphic LCD with easy-to-identify icons
  • Displays operational information, bump test, and calibration status
  • IntelliFlash for continuous visual confirmation of operation and compliance
  • Suitable for confined space entry
  • Compatible with MicroDock II automatic test and calibration system
  • Water resistant