Learn More
MSC BW Technologies by Honeywell GasAlertQuattro, Multi-Gas, 1 ea
Catalog No. 5000803287
Encompass
The GasAlertQuattro gas detector is a personal safety device designed to warn of hazardous gas at levels above user-defined alarm setpoints. It monitors 1, 2, 3, or 4 gases, including hydrogen sulfide (H2S), carbon monoxide (CO), combustible gases (LEL), and oxygen (O2).
  • Monitors 1, 2, 3, or 4 gases (H2S, CO, LEL, O2)
  • Warns of hazardous gas levels with visual, audio, and vibrate alarms
  • Liquid crystal display (LCD) shows readings, alarm setpoints, and messages
  • Single button operation for activation/deactivation and data viewing
  • Configurable via Fleet Manager II software
  • Performs battery test, segment test, and sensor self-test during startup
  • Supports rechargeable battery pack or 3 alkaline batteries
  • Designed for use in potentially explosive atmospheres
Catalog No. 50-008-03287 Supplier MSC Supplier No. 49069271
The GasAlertQuattro gas detector is a personal safety device designed to warn of hazardous gas at levels above user-defined alarm setpoints. It monitors 1, 2, 3, or 4 gases, including hydrogen sulfide (H2S), carbon monoxide (CO), combustible gases (LEL), and oxygen (O2).
  • Monitors 1, 2, 3, or 4 gases (H2S, CO, LEL, O2)
  • Warns of hazardous gas levels with visual, audio, and vibrate alarms
  • Liquid crystal display (LCD) shows readings, alarm setpoints, and messages
  • Single button operation for activation/deactivation and data viewing
  • Configurable via Fleet Manager II software
  • Performs battery test, segment test, and sensor self-test during startup
  • Supports rechargeable battery pack or 3 alkaline batteries
  • Designed for use in potentially explosive atmospheres