Learn More
ATCC BW5147.G.1.4; Lymphoma; Mouse (Mus musculus)
Catalog No. 502384581
Encompass
BW5147.G.1.4 is a t lymphoblast from the thymus of a mouse with lymphoma and can be used in immunology.
Catalog No. 50-238-4581 Supplier ATCC Supplier No. TIB48
BW5147.G.1.4 is a t lymphoblast from the thymus of a mouse with lymphoma and can be used in immunology.