Learn More
Being Scientific BWB-05 - 5L, Temperature Range:Amb+5℃ - 99℃
Catalog No. 5000217555
Encompass_Preferred
BEING Scientific General Purpose Laboratory Water Baths provide users with a constant temperature environment that only can be obtained when samples are totally immersed in water. Our line of lab water baths come in a variety of sizes and are suitable for direct heating and auxiliary heating of biological, chemical, physical, plant, chemical, and other experimental samples. Ideal applications for each BEING Water Bath include routine laboratory temperature control, E.Coli detection, sample thawing, bacterial detection, incubation microbial experiments, cell culture, and food testing pretreatment.
Catalog No. 50-002-17555 Supplier Being Scientific Supplier No. BW15005U
BEING Scientific General Purpose Laboratory Water Baths provide users with a constant temperature environment that only can be obtained when samples are totally immersed in water. Our line of lab water baths come in a variety of sizes and are suitable for direct heating and auxiliary heating of biological, chemical, physical, plant, chemical, and other experimental samples. Ideal applications for each BEING Water Bath include routine laboratory temperature control, E.Coli detection, sample thawing, bacterial detection, incubation microbial experiments, cell culture, and food testing pretreatment.