Learn More
Being Scientific BWZ-1 - 1.2 cu ft (33 Liters), Ambient+5 - 100℃, 17.1x12.5x9.5 (inch)435x317x250 (mm)
Catalog No. 5000217593
Encompass_Preferred
The BEING BWZ shaking water baths combine a general-purpose water bath with a reciprocating shaker motion to provide gentle stirring with precise laboratory temperature control suitable for direct and auxiliary heating of biological, chemical, physical, plant, chemical, and other experimental samples for Escherichia coli detection, sample thawing, bacterial detection, incubation microbial experiments, cell culture, food testing pretreatment, and more.
Catalog No. 50-002-17593 Supplier Being Scientific Supplier No. WZ21033U
The BEING BWZ shaking water baths combine a general-purpose water bath with a reciprocating shaker motion to provide gentle stirring with precise laboratory temperature control suitable for direct and auxiliary heating of biological, chemical, physical, plant, chemical, and other experimental samples for Escherichia coli detection, sample thawing, bacterial detection, incubation microbial experiments, cell culture, food testing pretreatment, and more.