Learn More
Cayman Chemical BX-795, 702675-74-9, 5 mg
SDP
Supplier:  Cayman Chemical 149325
Encompass_Preferred
Molecular Formula C23H26IN7O2S, Purity ≥98%, Formula Weight 591.5, Synonyms: N-[3-[[5-iodo-4-[[3-[(2-thienylcarbonyl)amino]propyl]amino]-2-pyrimidinyl]amino]phenyl]-1-pyrrolidinecarboxamide; A potent, ATP-competitive inhibitor of PDK1 in vitro (IC50 = 11 nM) and in cells (IC50 = 300 nM); also inhibits ERK8, MNK2, Aurora B, Aurora C, MARKs 1-4,TBK1, IKK, and additional kinases at comparable concentrations.
Catalog No. NC0685258