Learn More
Selleck Chemical LLC BX-912 S1275-2mg
SDP
Catalog No. 503108348
Encompass_Preferred
BX912 is a potent and specific PDK1 inhibitor with IC50 of 12 nM 9- and 105- fold greater selectivity for PDK1 than PKA and PKC in cell-free assays respectively In comparison to GSK3 selectivity for PDK1 is 600-fold
Catalog No. 50-310-8348 Supplier Selleck Chemical LLC Supplier No. S12752MG
BX912 is a potent and specific PDK1 inhibitor with IC50 of 12 nM 9- and 105- fold greater selectivity for PDK1 than PKA and PKC in cell-free assays respectively In comparison to GSK3 selectivity for PDK1 is 600-fold