Learn More
Medchemexpress LLC HY-11005 5mg Medchemexpress, BX-912 CAS:702674-56-4 Purity:>98%
SDP
Catalog No. 501968737
Encompass_Preferred
Medchemexpress, HY-11005 5mg BX-912 CAS:702674-56-4 BX-912 is a direct, selective, and ATP-competitive PDK1 inhibitor (IC50=26 nM). BX-912 blocks PDK1/Akt signaling in tumor cells and inhibits the anchorage-dependent growth of a variety of tumor cell lines in culture or induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8737 Supplier Medchemexpress LLC Supplier No. HY110055MG
Medchemexpress, HY-11005 5mg BX-912 CAS:702674-56-4 BX-912 is a direct, selective, and ATP-competitive PDK1 inhibitor (IC50=26 nM). BX-912 blocks PDK1/Akt signaling in tumor cells and inhibits the anchorage-dependent growth of a variety of tumor cell lines in culture or induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.