Learn More
Biotage BX PHYTIP COL W/PROPLUS AFF RS
SDP
Catalog No. NC1859750
Encompass
Box of 96 PhyTip columns (1 ml volume) containing 80 μl of ProPlus affinity resin per column
Catalog No. NC1859750 Supplier Biotage Supplier No. PTH918007
Box of 96 PhyTip columns (1 ml volume) containing 80 μl of ProPlus affinity resin per column