Learn More
MSC BX SAFETY EYEWEAR
Catalog No. 5000803453
Encompass
3M BX Readers Magnifying Safety Glasses combine eye protection with reading enhancement. They feature polycarbonate lenses that absorb 99.9% UVA and UVB up to 380 nm, making them suitable for various laboratory and industrial applications requiring vision correction.
  • Sleek, contoured design combines style and function
  • Magnifying +1.5 diopters combine eye protection with reading enhancement
  • Soft nose bridge offers pleasant extended wear
  • Adjustable length temples and lens angle allow for customized fit
  • Anti-fog coatings improve visibility under hot and humid conditions
  • Polycarbonate lenses absorb 99.9% UVA and UVB up to 380 nm
Catalog No. 50-008-03453 Supplier MSC Supplier No. 06136808
3M BX Readers Magnifying Safety Glasses combine eye protection with reading enhancement. They feature polycarbonate lenses that absorb 99.9% UVA and UVB up to 380 nm, making them suitable for various laboratory and industrial applications requiring vision correction.
  • Sleek, contoured design combines style and function
  • Magnifying +1.5 diopters combine eye protection with reading enhancement
  • Soft nose bridge offers pleasant extended wear
  • Adjustable length temples and lens angle allow for customized fit
  • Anti-fog coatings improve visibility under hot and humid conditions
  • Polycarbonate lenses absorb 99.9% UVA and UVB up to 380 nm