Learn More
MSC BX SAFETY EYEWEAR
Catalog No. 5000782427
Encompass
The 3M BX Reader Protective Eyewear features a silver/black frame and gray anti-fog lens. These safety glasses are designed for workers who have difficulty reading small print or are engaged in detailed work. The polycarbonate lenses absorb 99.9% UVA and UVB up to 380 nm and meet ANSI Z87.1 and CSA Z94.3 standards.
  • Sleek, contoured design
  • Magnifying +2.5 diopter for reading enhancement
  • Adjustable length temples and lens angle for customized fit
  • Polycarbonate lenses protect eyes from impact and UV rays
  • Soft adjustable nose bridge
Catalog No. 50-007-82427 Supplier MSC Supplier No. 06136824
The 3M BX Reader Protective Eyewear features a silver/black frame and gray anti-fog lens. These safety glasses are designed for workers who have difficulty reading small print or are engaged in detailed work. The polycarbonate lenses absorb 99.9% UVA and UVB up to 380 nm and meet ANSI Z87.1 and CSA Z94.3 standards.
  • Sleek, contoured design
  • Magnifying +2.5 diopter for reading enhancement
  • Adjustable length temples and lens angle for customized fit
  • Polycarbonate lenses protect eyes from impact and UV rays
  • Soft adjustable nose bridge