Learn More
MSC BX SAFETY EYEWEAR
Catalog No. 5000767683
Encompass
3M Magnifying Safety Glasses offer integrated eye protection and reading enhancement. They feature +2.50 clear, anti-fog coated polycarbonate lenses that absorb 99.9% UVA and UVB up to 380 nm. Designed for comfort and a customized fit, these glasses include a soft adjustable nose bridge and adjustable temples. They comply with ANSI Z87.1 and CSA Z94.3 standards.
  • Sleek, contoured design for style and function
  • Integrates eye protection with reading enhancement (+2.50 diopter)
  • Soft adjustable nose bridge for pleasant extended wear
  • Adjustable length temples and lens angle for customized fit
  • Polycarbonate lenses absorb 99.9% UVA and UVB up to 380 nm
  • Anti-fog coating improves visibility in hot and humid conditions
  • Meets ANSI Z87.1 and CSA Z94.3 safety standards
Catalog No. 50-007-67683 Supplier MSC Supplier No. 06136790
3M Magnifying Safety Glasses offer integrated eye protection and reading enhancement. They feature +2.50 clear, anti-fog coated polycarbonate lenses that absorb 99.9% UVA and UVB up to 380 nm. Designed for comfort and a customized fit, these glasses include a soft adjustable nose bridge and adjustable temples. They comply with ANSI Z87.1 and CSA Z94.3 standards.
  • Sleek, contoured design for style and function
  • Integrates eye protection with reading enhancement (+2.50 diopter)
  • Soft adjustable nose bridge for pleasant extended wear
  • Adjustable length temples and lens angle for customized fit
  • Polycarbonate lenses absorb 99.9% UVA and UVB up to 380 nm
  • Anti-fog coating improves visibility in hot and humid conditions
  • Meets ANSI Z87.1 and CSA Z94.3 safety standards