Learn More
Medchemexpress LLC Bx430 | 688309-70-8 | 99.2% | 413.11 | 10 MG
SDP
Catalog No. 5000343963
Encompass_Preferred
BX430 is a potent and selective noncompetitive allosteric human P2X4 receptor channels antagonist with an IC50 of 0.54 μM. It exhibits species specificity and is used for chronic pain and cardiovascular disease.
  • Potent and selective noncompetitive allosteric human P2X4 receptor channels antagonist
  • Exhibits species specificity
  • Used for chronic pain and cardiovascular disease
  • For research use only
Catalog No. 50-003-43963 Supplier Medchemexpress LLC Supplier No. HY110237100MG
BX430 is a potent and selective noncompetitive allosteric human P2X4 receptor channels antagonist with an IC50 of 0.54 μM. It exhibits species specificity and is used for chronic pain and cardiovascular disease.
  • Potent and selective noncompetitive allosteric human P2X4 receptor channels antagonist
  • Exhibits species specificity
  • Used for chronic pain and cardiovascular disease
  • For research use only