Learn More
Medchemexpress LLC Bx430 | 688309-70-8 | 99.2% | 413.11 | 50MG
SDP
Catalog No. 5000343868
Encompass_Preferred
BX430 is a potent and selective noncompetitive allosteric human P2X4 receptor channels antagonist with an IC50 of 0.54 μM. It exhibits species specificity and is utilized for chronic pain and cardiovascular disease.
  • Potent and selective noncompetitive allosteric human P2X4 receptor channels antagonist.
  • IC50 of 0.54 μM against P2X4 receptor channels.
  • Exhibits species specificity.
  • Has virtually no functional impact on other P2X subtypes (P2X1-P2X3, P2X5, and P2X7).
  • Potent antagonist of zebrafish P2X4 but no effect on rat and mouse P2X4 orthologs.
  • Used for chronic pain and cardiovascular disease.
Catalog No. 50-003-43868 Supplier Medchemexpress LLC Supplier No. HY11023750MG
BX430 is a potent and selective noncompetitive allosteric human P2X4 receptor channels antagonist with an IC50 of 0.54 μM. It exhibits species specificity and is utilized for chronic pain and cardiovascular disease.
  • Potent and selective noncompetitive allosteric human P2X4 receptor channels antagonist.
  • IC50 of 0.54 μM against P2X4 receptor channels.
  • Exhibits species specificity.
  • Has virtually no functional impact on other P2X subtypes (P2X1-P2X3, P2X5, and P2X7).
  • Potent antagonist of zebrafish P2X4 but no effect on rat and mouse P2X4 orthologs.
  • Used for chronic pain and cardiovascular disease.