Learn More
Selleck Chemical LLC BX471-5mg
SDP
Supplier:  Selleck Chemical LLC S76045MG
Encompass_Preferred
BX471 (ZK811752, BAY 865047, SH T 04268H) is a potent nonpeptide CC chemokine receptor-1 (CCR1) antagonist with Ki values of 1 nM and 5.5 nM in both MIP-1α and MCP-3 binding to CCR1-transfected HEK293 cells, respectively.
Catalog No. 50-238-9951