Learn More
Medchemexpress LLC HY-12080A 10mg Medchemexpress, BX471 (hydrochloride) CAS:288262-96-4 Purity:>98%
SDP
Catalog No. 501969614
Encompass_Preferred
Medchemexpress, HY-12080A 10mg BX471 (HCl) CAS:288262-96-4 BX471 hydrochloride (ZK-811752 hydrochloride) is a potent, selective non-peptide CCR1 antagonist with Ki of 1 nM for human CCR1, and exhibits 250-fold selectivity for CCR1 over CCR2, CCR5 and CXCR4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9614 Supplier Medchemexpress LLC Supplier No. HY12080A10MG
Medchemexpress, HY-12080A 10mg BX471 (HCl) CAS:288262-96-4 BX471 hydrochloride (ZK-811752 hydrochloride) is a potent, selective non-peptide CCR1 antagonist with Ki of 1 nM for human CCR1, and exhibits 250-fold selectivity for CCR1 over CCR2, CCR5 and CXCR4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.