Learn More
Medchemexpress LLC Bx471 hydrochloride | 288262-96-4 | 99.9% | 1 ML
SDP
Catalog No. 5000230834
Encompass_Preferred
BX471 hydrochloride (ZK-811752 hydrochloride) is a potent, selective, non-peptide CCR1 antagonist (Ki = 1 nM) that shows approximately 250-fold selectivity over CCR2, CCR5, and CXCR4. It is supplied for research as a ready-to-use 10 mM solution in DMSO (1 mL) or as solid quantities for reconstitution, and is commonly used in studies of CCR1 pharmacology and inflammation.
  • Potent CCR1 antagonist with Ki = 1 nM.
  • Approximately 250-fold selectivity versus CCR2, CCR5, and CXCR4.
  • Supplied as a 10 mM solution in DMSO (1 mL) or as solids for reconstitution.
  • High purity suitable for research applications.
Catalog No. 50-002-30834 Supplier Medchemexpress LLC Supplier No. HY12080A10MM 1ML
BX471 hydrochloride (ZK-811752 hydrochloride) is a potent, selective, non-peptide CCR1 antagonist (Ki = 1 nM) that shows approximately 250-fold selectivity over CCR2, CCR5, and CXCR4. It is supplied for research as a ready-to-use 10 mM solution in DMSO (1 mL) or as solid quantities for reconstitution, and is commonly used in studies of CCR1 pharmacology and inflammation.
  • Potent CCR1 antagonist with Ki = 1 nM.
  • Approximately 250-fold selectivity versus CCR2, CCR5, and CXCR4.
  • Supplied as a 10 mM solution in DMSO (1 mL) or as solids for reconstitution.
  • High purity suitable for research applications.